Learn More
AdipoGen 1-Carboxymethylmetronidazole
SDP
Catalog No. 502560528
Encompass_Preferred
Chemical. CAS 1010-93-1. Formula C6H7N3O4. MW 185.1. Synthetic. A main acid metabolite of the antibiotic metronidazole showing some antibacterial activity. Compound can be used as analytical reference material.
Catalog No. 50-256-0528 Supplier AdipoGen Supplier No. CDXM0091M100
Chemical. CAS 1010-93-1. Formula C6H7N3O4. MW 185.1. Synthetic. A main acid metabolite of the antibiotic metronidazole showing some antibacterial activity. Compound can be used as analytical reference material.