Learn More
Alkali Scientific 1" Cardboard Freezer Boxes, fits 0.2mL PCR tubes with 196 place divider, 100/Case
SDP
Catalog No. 5000662868
Encompass_Preferred
Utilized to organize and store your 0.2mL and 0.1mL PCR 8 strips and 0.2mL & 0.1mL single tubes. Fits both flat capped and round cap 0.2mL/0.1mL tubes in both 12 strip and 8 strip & single tube format. The supplied grid is 196 (14 x 14) inner compartments, thus fitting for example, 24 x 8 strip tubes with 4 compartments remaining open. These economical cryo/freezer storage boxes offer excellent low temperature storage. Moisture repellent white plasticized coating protects boxes from effects of liquid nitrogen or condensation. Our 1" boxes arrive with 14 x 14 place dividers already within. You can double stack these within 2" stainless steel storage rack compartments. Snug fitting lid prevents accidental separation if dropped. Use in mechanical freezers or liquid nitrogen tanks to –196°C Constructed of sturdy cardboard with moisture repellent coating Designed with small holes on the bottom to allow for drainage. Designed to hold 196 PCR tubes with a max outer diameter of 8 mm.
Catalog No. 50-006-62868 Supplier Alkali Scientific Supplier No. FS196C100
Utilized to organize and store your 0.2mL and 0.1mL PCR 8 strips and 0.2mL & 0.1mL single tubes. Fits both flat capped and round cap 0.2mL/0.1mL tubes in both 12 strip and 8 strip & single tube format. The supplied grid is 196 (14 x 14) inner compartments, thus fitting for example, 24 x 8 strip tubes with 4 compartments remaining open. These economical cryo/freezer storage boxes offer excellent low temperature storage. Moisture repellent white plasticized coating protects boxes from effects of liquid nitrogen or condensation. Our 1" boxes arrive with 14 x 14 place dividers already within. You can double stack these within 2" stainless steel storage rack compartments. Snug fitting lid prevents accidental separation if dropped. Use in mechanical freezers or liquid nitrogen tanks to –196°C Constructed of sturdy cardboard with moisture repellent coating Designed with small holes on the bottom to allow for drainage. Designed to hold 196 PCR tubes with a max outer diameter of 8 mm.