Learn More
Bioss 1-Catenin - central region An
SDP
Catalog No. 5000065718
Encompass_Preferred
1-Catenin (central region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB IP M354Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65718 Supplier Bioss Supplier No. BSM70388M
1-Catenin (central region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB IP M354Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol