Learn More
Bioss 1-Catenin - Tyr-228 Phosphos
SDP
Catalog No. 5000065407
Encompass_Preferred
1-Catenin (Tyr-228) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M356Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65407 Supplier Bioss Supplier No. BSM70389M
1-Catenin (Tyr-228) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M356Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol