Learn More
Waters Corp Cartridge Stand, 1 cc, 96-Well Plate Format, Gravity Flow
Catalog No. 502391123 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This cartridge stand is designed to properly hold cartridges for gravity-assisted flow, facilitating desalting and buffer exchanging of proteins. It features a 96-well plate format, making it suitable for both manual and automated workflows.
  • Facilitates gravity-assisted flow for desalting and buffer exchanging proteins.
  • Compatible with 96-well plate format.
  • Suitable for manual workflows using an 8-channel pipette.
  • Amenable to automated workflows.
  • Includes one 1 cc cartridge stand.
Catalog No. 50-239-1123 Supplier Waters Corp Supplier No. 186010128
This cartridge stand is designed to properly hold cartridges for gravity-assisted flow, facilitating desalting and buffer exchanging of proteins. It features a 96-well plate format, making it suitable for both manual and automated workflows.
  • Facilitates gravity-assisted flow for desalting and buffer exchanging proteins.
  • Compatible with 96-well plate format.
  • Suitable for manual workflows using an 8-channel pipette.
  • Amenable to automated workflows.
  • Includes one 1 cc cartridge stand.