Learn More
Terumo™ 1 cc Luer Slip Syringes and Conventional Needles
Catalog No. 22290021 Shop All Terumo Medical Corporation Products
Change view
Click to view available options
Needle Gauge:
25 G
26 G
27 G
Includes:
Syringe Only
Syringe and Needle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Includes
22-290-021 27 G Syringe and Needle
22-290-018 - Syringe Only
22-290-019 25 G Syringe and Needle
22-290-020 26 G Syringe and Needle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-290-021 Supplier Terumo™ Supplier No. SS01T2713
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
1 cc Luer Slip Syringes and Conventional Needles are available with various gauge and length needles.
  • Superior sharpness and smoothness
  • Ultra Sharp Needles
  • Clear Barrels
  • Bold, Precise Scale Markings
  • Smooth Plunger Action
  • Tamper Evident Peel Packs
Specifications
Material Plastic
Color Clear
Needle Gauge 27 G
Includes Syringe and Needle
Length (English) Needle 1/2 in.
Product Type Luer Slip Syringe
Disposable Single-use
Graduations 0.01 cc
Quantity 100 Pk., 10 Pk./Cs.
Volume (Metric) 1 mL
Connection Type Luer Slip
Graduated Yes
Sterility Sterile