Learn More
Thermo Scientific™ Cartridges for Multidrop™ Pico Digital Dispensers
GREENER_CHOICE
Catalog No. NC1789360 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
1 nL to 10 μL
11 pL to 10 μL
Channels:
1
4
8
Model:
1-channel 200 μL Cartridges
1-channel 20 μL Cartridges
4-channel 200 μL Standard Cartridges
8-channel 20 μL Standard Cartridges
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Channels Model
NC1789360 11 pL to 10 μL 8 8-channel 20 μL Standard Cartridges
NC1789352 11 pL to 10 μL 1 1-channel 20 μL Cartridges
NC1789355 1 nL to 10 μL 1 1-channel 200 μL Cartridges
NC1789357 1 nL to 10 μL 4 4-channel 200 μL Standard Cartridges
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. NC1789360 Supplier Thermo Scientific™ Supplier No. LTR0004
Cartridges for Multidrop™ Pico Digital Dispensers allow dispensing of one to eight fluids per cartridge, from 8 pL to 10 μL of DMSO, aqueous fluids, or master mixes, with or without small molecules or biomolecules.
The cartridges are designed for use with Multidrop™ Pico Digital Dispensers, dispensing volumes from 8 pL to 10 μL. Use the 1-channel cartridges with the Multidrop™ Pico 1, and the 4- or 8-channel standard or surfactant free cartridges with the Multidrop™ Pico 8 Surfactant Free (Pico 8 sf).
Features:
  • Single-Channel Cartridges: Use the 1-channel cartridges with the Multidrop™ Pico 1 for single fluid dispensing at a time.
  • Multi-Channel Cartridges: Use the 4- or 8-channel cartridges with the Multidrop™ Pico 8 sf to dispense multiple fluids per cartridge.
  • Surfactant Free Cartridges: The Multidrop™ Pico 8 sf with the surfactant free cartridges can be used to dispense aqueous fluids without surfactant.
  • Contamination-Free Dispensing: Non-contact dispensing and single-use cartridges prevent carry-over and contamination.
  • Fluid Compatibility: Ideal for DMSO-based fluids, aqueous fluids, or master mixes, with or without small molecules or biomolecules. Aqueous fluids can be dispensed without surfactant when using the surfactant free cartridges with the Multidrop™ Pico 8 sf.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Digital Dispenser Cartridge
Channels 8
Volume (Metric) 11 pL to 10 μL
For Use With (Equipment) Multidrop Pico 8 Digital Dispenser, Multidrop Pico 8 Surfactant Free Digital Dispenser
Model 8-channel 20 μL Standard Cartridges
No. per Pack Case of 20