Learn More
Instech Vascular Access Button™ for Mouse, Magnetic, 1 Channel 22ga, with Injector
SDP
Catalog No. NC2816919
Encompass
One channel Vascular Access Button™ for mice, 14mm polyester felt, the standard configuration; tissue will grow into the felt over time, securing it in place. 22 ga port catheter connector. Injector included. The mouse VAB™ is a transcutaneous button that permits quick, aseptic access to a catheterized vessel either directly with a syringe or outside the cage via a tether.
Catalog No. NC2816919 Supplier Instech Supplier No. VABM1B/22
One channel Vascular Access Button™ for mice, 14mm polyester felt, the standard configuration; tissue will grow into the felt over time, securing it in place. 22 ga port catheter connector. Injector included. The mouse VAB™ is a transcutaneous button that permits quick, aseptic access to a catheterized vessel either directly with a syringe or outside the cage via a tether.