Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2,3-dimethoxybenzene 97.0+%, TCI America™
SDP

Catalog No. C13555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13555G 5 g
C13551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13555G Supplier TCI America Supplier No. C13555G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90282-99-8
Molecular Formula C8H9ClO2
Molecular Weight (g/mol) 172.608
MDL Number MFCD00191399
InChI Key JVCXXMPHTZGGNV-UHFFFAOYSA-N
PubChem CID 56060
IUPAC Name 1-chloro-2,3-dimethoxybenzene
SMILES COC1=C(C(=CC=C1)Cl)OC

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 98°C
Formula Weight 172.61
Percent Purity ≥97.0% (GC)
Chemical Name or Material 1-Chloro-2,3-dimethoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.