Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2,4-dinitrobenzene, 98%

Catalog No. AAA137740E
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA137740E 2500 g
AAA1377422 100 g
AAA1377436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA137740E Supplier Thermo Scientific Chemicals Supplier No. A137740E
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 97-00-7 | C6H3ClN2O4 | 202.55 g/mol

1-Chloro-2,4-dinitrobenzene is used as a reagent for the detection and determination of pyridine compounds. It has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH). It is an irreversible inhibitor of human thioredoxin reductase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2,4-dinitrobenzene is used as a reagent for the detection and determination of pyridine compounds. It has been used as alkylating agent to evaluate the depletion of intracellular erythrocyte glutathione (GSH). It is an irreversible inhibitor of human thioredoxin reductase.

Solubility
Soluble in chloroform, ether, hot alcohol, and benzene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, ammonia. Reacts with hydrazine hydrate.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-00-7
Molecular Formula C6H3ClN2O4
Molecular Weight (g/mol) 202.55
MDL Number MFCD00007075
InChI Key VYZAHLCBVHPDDF-UHFFFAOYSA-N
Synonym 2,4-dinitrochlorobenzene, dinitrochlorobenzene, dncb, cdnb, chlorodinitrobenzene, 4-chloro-1,3-dinitrobenzene, 2,4-dinitrophenyl chloride, benzene, 1-chloro-2,4-dinitro, dinitrochlorobenzol, 1,3-dinitro-4-chlorobenzene
PubChem CID 6
ChEBI CHEBI:34718
IUPAC Name 1-chloro-2,4-dinitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point 48°C to 52°C
Density 1.7
Boiling Point 314°C to 316°C
Flash Point 186°C (366°F)
Quantity 2500 g
UN Number UN3441
Beilstein 613161
Merck Index 14,2136
Solubility Information Soluble in chloroform,ether,hot alcohol,and benzene. Insoluble in water.
Formula Weight 202.55
Percent Purity 98%
Chemical Name or Material 1-Chloro-2,4-dinitrobenzene
Show More Show Less
Hazard Category H302-H310-H315-H317-H318
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P261-P262-P264b-P270-P272-P280-P301+P312-P302+P350-P305+P351+P338-P310-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORODINITROBENZENES, SOLID
EINECSNumber 202-551-4
RTECSNumber CZ0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.