Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2,5-diethoxybenzene 98.0+%, TCI America™
SDP

Catalog No. C1576500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1576500G 500 g
C157625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1576500G Supplier TCI America Supplier No. C1576500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 52196-74-4
Molecular Formula C10H13ClO2
Molecular Weight (g/mol) 200.662
MDL Number MFCD00026765
InChI Key ZIMKMIAIVORSSX-UHFFFAOYSA-N
Synonym 2,5-Diethoxychlorobenzene
PubChem CID 40273
IUPAC Name 2-chloro-1,4-diethoxybenzene
SMILES CCOC1=CC(=C(C=C1)OCC)Cl

Specifications

Color Yellow
Quantity 500 g
Physical Form Clear Liquid at 20°C
Boiling Point 266°C
Formula Weight 200.66
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1-Chloro-2,5-diethoxybenzene
EINECSNumber (3)-0919
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.