Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-chloro-2-(ethylsulfonyl)ethane

Catalog No. CDS011744 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS011744 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS011744 Supplier Sigma Aldrich Supplier No. CDS011744100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,2-dimethylhexane is one of the products of the corona discharge of a simulated titanαs atmosphere (2% argon in nitrogen and 10% methane) at 500 torr and 298 k (using GC-ftir-ms techniques)
  • It was used to study the adsorption of linear and branched alkanes on a microporous silicoaluminophosphate material (sapo-5) using chromatographic techniques

Specifications

CAS 25027-40-1
Molecular Formula C4H9ClO2S
Linear Formula C4H9ClO2S
MDL Number MFCD01752806
Quantity 1 g
Molecular Weight (g/mol) 156.63
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.