Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2-methyl-2-phenylpropane 98.0+%, TCI America™
SDP

Catalog No. C1068500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1068500ML 500 mL
C106825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1068500ML Supplier TCI America Supplier No. C1068500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 515-40-2
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.66
MDL Number MFCD00000940
InChI Key DNXXUUPUQXSUFH-UHFFFAOYSA-N
Synonym neophyl chloride, 1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, benzene, 2-chloro-1,1-dimethylethyl, 2-methyl-2-phenylpropyl chloride, neophylchloride, 2-chloro-1,1-dimethylethyl benzene, unii-of6e0410nf, beta-chloro-tert-butylbenzene, 2-chloromethyl-2-phenylpropane
PubChem CID 68191
IUPAC Name (1-chloro-2-methylpropan-2-yl)benzene
SMILES CC(C)(CCl)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 222°C
Quantity 500 mL
Formula Weight 168.66
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-2-methyl-2-phenylpropane
EINECSNumber (3)-3950
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.