Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2-methylpropane 98%, Thermo Scientific™

Catalog No. AC176202500
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC176202500 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC176202500 Supplier Thermo Scientific Chemicals Supplier No. 176202500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-36-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
InChI Key QTBFPMKWQKYFLR-UHFFFAOYSA-N
Synonym isobutyl chloride, propane, 1-chloro-2-methyl, 1-chloro-2-methyl-propane, isobutylchlorid, iso-c4h9cl, sjphadikup@, acmc-1akdy, propane,1-chloro-2-methyl, ksc491k9j, 1-chloro-2-methylpropane
PubChem CID 10554
IUPAC Name 1-chloro-2-methylpropane
SMILES CC(C)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Linear Formula (CH3)2CHCH2Cl
Quantity 250 mL
Beilstein 01, 124
Merck Index 15, 5183
Formula Weight 92.57
Chemical Name or Material 1-Chloro-2-methylpropane, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.