Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2-methylpropyl chloroformate, 98%

Catalog No. AC314470010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314470010 1 g Glass bottle
AC314470050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC314470010 Supplier Thermo Scientific Chemicals Supplier No. 314470010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 92600-11-8 | C5H8Cl2O2 | 171.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92600-11-8
Molecular Formula C5H8Cl2O2
Molecular Weight (g/mol) 171.02
MDL Number MFCD00012387
InChI Key BOGLZTPABVHWDD-BYPYZUCNSA-N
Synonym 1-chloro-2-methylpropyl chloroformate, acmc-20aow3, 1-chloro-2-methylpropylchloroformate, 1-chloro-2-methylpropyl carbonochloridate, chloro 1-chloro-2-methylpropoxy methanone, carbonochloridic acid,1-chloro-2-methylpropyl ester
PubChem CID 2733326
SMILES CC(C)[C@@H](Cl)OC(Cl)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1700g/mL
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4310 to 1.433
Linear Formula ClCO2CH(Cl)CH(CH3)2
Quantity 1 g
Specific Gravity 1.17
IUPAC Name (1R)-1-chloro-2-methylpropyl carbonochloridate
Formula Weight 171.02
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-chloro-2-methylpropyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.