Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2-(trifluoromethoxy)benzene 98.0+%, TCI America™
SDP

Catalog No. C209125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C209125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C209125G Supplier TCI America Supplier No. C209125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 450-96-4
Molecular Formula C7H4ClF3O
Molecular Weight (g/mol) 196.55
MDL Number MFCD03840206
InChI Key NZRLCYJRHKUVCV-UHFFFAOYSA-N
PubChem CID 2782782
IUPAC Name 1-chloro-2-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=CC=C1Cl

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 1993
Boiling Point 143°C
Formula Weight 196.55
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1-Chloro-2-(trifluoromethoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.