Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-3,5-difluorobenzene 97.0+%, TCI America™
SDP

Catalog No. C25145G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25145G 5 g
C251425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25145G Supplier TCI America Supplier No. C25145G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1435-43-4
Molecular Formula C6H3ClF2
Molecular Weight (g/mol) 148.537
MDL Number MFCD00041518
InChI Key RFKBODCWHNDUTJ-UHFFFAOYSA-N
PubChem CID 137000
IUPAC Name 1-chloro-3,5-difluorobenzene
SMILES C1=C(C=C(C=C1F)Cl)F

Specifications

Color Yellow
Boiling Point 116°C
Quantity 5 g
UN Number 1993
Synonym 3,5-difluorochlorobenzene, benzene, 1-chloro-3,5-difluoro, benzene,1-chloro-3,5-difluoro, 1-chloro-3,5-difluoro-benzene, 3,5-difluorochlorobenzene, 5-difluorochlorobenzene, pubchem4318, 3,5-difluorchlorbenzol, acmc-209cqr, intermediates-zcf02092, 1-chloro-3,5difluorobenzene
Formula Weight 148.54
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-3,5-difluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.