Learn More
Medchemexpress LLC 1-chloro-3,5-dimethoxybenzene | 7051-16-3 | MFCD00008382 | 98.3% | 172.61 g/mol | C8H9ClO2 | 10 G
SDP
Catalog No. 5000300649
Encompass_Preferred
5-Chlororesorcinol dimethyl ether (1-chloro-3,5-dimethoxybenzene) is a chlorinated resorcinol derivative used as a synthetic intermediate in pharmaceutical and organic synthesis. It is supplied as a high-purity, well-characterized material suitable for laboratory-scale synthesis and analytical work.
  • High purity verified by GC (98.34%).
  • Molecular weight 172.61 g/mol and formula C8H9ClO2.
  • Suitable as a building block for pharmaceutical intermediates.
  • Characterized by 1H NMR and low water content (0.03% KF).
  • Stable solid under recommended storage conditions.
Catalog No. 50-003-00649 Supplier Medchemexpress LLC Supplier No. HYW01685910G
5-Chlororesorcinol dimethyl ether (1-chloro-3,5-dimethoxybenzene) is a chlorinated resorcinol derivative used as a synthetic intermediate in pharmaceutical and organic synthesis. It is supplied as a high-purity, well-characterized material suitable for laboratory-scale synthesis and analytical work.
  • High purity verified by GC (98.34%).
  • Molecular weight 172.61 g/mol and formula C8H9ClO2.
  • Suitable as a building block for pharmaceutical intermediates.
  • Characterized by 1H NMR and low water content (0.03% KF).
  • Stable solid under recommended storage conditions.