Learn More
Medchemexpress LLC 1-chloro-3,5-dimethoxybenzene | 7051-16-3 | MFCD00008382 | 98.3% | 172.61 g·mol⁻¹ | C8H9ClO2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW016859
Encompass_Preferred
5-Chlororesorcinol dimethyl ether (1-chloro-3,5-dimethoxybenzene; CAS 7051-16-3) is a small-molecule synthetic intermediate commonly used in medicinal chemistry and organic synthesis. Supplied for research use, it is typically provided as a solid with reported purity around 98% and is used as a building block in multi-step syntheses and library preparation.
  • High purity suitable for synthesis and analytical characterization.
  • Solid form with standard room-temperature storage recommendations.
  • Commonly used as a building block in pharmaceutical and medicinal chemistry syntheses.
  • Offered in multiple package sizes for research-scale work.
  • Molecular formula C8H9ClO2 and molecular weight 172.61 g·mol⁻¹.
Catalog No. 50-003-03821