Learn More
Medchemexpress LLC 1-chloro-3,5-dimethoxybenzene | 7051-16-3 | MFCD00008382 | 98.3% | 172.61 g/mol | C8H9ClO2 | 50 G
SDP
Catalog No. 5000300429
Encompass_Preferred
5-Chlororesorcinol dimethyl ether (1-chloro-3,5-dimethoxybenzene) is a chlorinated dimethoxybenzene used as a synthetic intermediate in pharmaceutical and fine-chemical synthesis. Supplied for research applications, it functions as a building block for preparation and modification of aromatic compounds.
  • High purity (≈98.3%) suitable for laboratory synthesis
  • Light yellow solid near room temperature; melting/transition ≈34-36 °C
  • Molecular formula C8H9ClO2; molecular weight ≈172.61 g/mol
  • Offered in common lab pack sizes for small-scale synthesis
  • Intended for research use as a synthetic intermediate
Catalog No. 50-003-00429 Supplier Medchemexpress LLC Supplier No. HYW01685950G
5-Chlororesorcinol dimethyl ether (1-chloro-3,5-dimethoxybenzene) is a chlorinated dimethoxybenzene used as a synthetic intermediate in pharmaceutical and fine-chemical synthesis. Supplied for research applications, it functions as a building block for preparation and modification of aromatic compounds.
  • High purity (≈98.3%) suitable for laboratory synthesis
  • Light yellow solid near room temperature; melting/transition ≈34-36 °C
  • Molecular formula C8H9ClO2; molecular weight ≈172.61 g/mol
  • Offered in common lab pack sizes for small-scale synthesis
  • Intended for research use as a synthetic intermediate