Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-3-fluorobenzene, 99%

Catalog No. AAA1312314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1312314 25 g
AAA1312322 100 g
AAA1312336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1312314 Supplier Thermo Scientific Chemicals Supplier No. A1312314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 625-98-9 | C6H4ClF | 130.55 g/mol

1-Chloro-3-fluorobenzene used as medicine and liquid crystal intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3-fluorobenzene used as medicine and liquid crystal intermediates.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-98-9
Molecular Formula C6H4ClF
Molecular Weight (g/mol) 130.55
MDL Number MFCD00000569
InChI Key VZHJIJZEOCBKRA-UHFFFAOYSA-N
Synonym m-chlorofluorobenzene, 3-chlorofluorobenzene, benzene, 1-chloro-3-fluoro, m-fluorochlorobenzene, 3-fluorochlorobenzene, 1-fluoro-3-chlorobenzene, 1-chloro-3-fluoro-benzene, m-fluoro chlorobenzene, pubchem3419, 3-fluoro-chlorobenzene
PubChem CID 223087
IUPAC Name 1-chloro-3-fluorobenzene
SMILES FC1=CC=CC(Cl)=C1

Specifications

Density 1.224
Boiling Point 126°C to 128°C
Flash Point 20°C (68°F)
Refractive Index 1.494
Quantity 25 g
UN Number UN1993
Beilstein 2039303
Solubility Information Not miscible in water.
Formula Weight 130.55
Percent Purity 99%
Chemical Name or Material 1-Chloro-3-fluorobenzene
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H332-H315-H319-H335
Highly flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 210-919-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.