Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-4-fluorobenzene 95.0+%, TCI America™
SDP

Catalog No. C065925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C065925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C065925G Supplier TCI America Supplier No. C065925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 352-33-0
Melting Point -26°C
Color Yellow
Boiling Point 130°C
Molecular Formula C6H4ClF
Quantity 25 g
MDL Number MFCD00000603
UN Number 1993
Synonym p-chlorofluorobenzene, p-fluorochlorobenzene, 4-chlorofluorobenzene, benzene, 1-chloro-4-fluoro, 1-fluoro-4-chlorobenzene, 1-chloro-4-fluoro-benzene, 4-fluorochlorobenzene, 1,4-fluorochlorobenzene, p-chloro-fluorobenzene, pubchem3420
InChI Key RJCGZNCCVKIBHO-UHFFFAOYSA-N
SMILES FC1=CC=C(Cl)C=C1
IUPAC Name 1-chloro-4-fluorobenzene
Molecular Weight (g/mol) 130.55
PubChem CID 9604
Formula Weight 130.55
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-4-fluorobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.