Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-9,10-bis(phenylethynyl)anthracene 96.0+%, TCI America™
SDP

Catalog No. C128910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C128910G 10 g
C12891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C128910G Supplier TCI America Supplier No. C128910G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41105-35-5
Molecular Formula C30H17Cl
Molecular Weight (g/mol) 412.92
MDL Number MFCD00012047
InChI Key IMMCAKJISYGPDQ-UHFFFAOYSA-N
Synonym 1-chloro-9,10-bis phenylethynyl anthracene, 1-chloro-bpea, anthracene, 1-chloro-9,10-bis phenylethynyl, 1-chloro-9,10-bis 2-phenylethynyl anthracene, anthracene, 9,10-bis phenylethynyl-1-chloro, anthracene, 1-chloro-9,10-bis 2-phenylethynyl, 1-chloro-bpee, cbpea, 1-cl-bpee, acmc-1an87
PubChem CID 170465
IUPAC Name 1-chloro-9,10-bis(2-phenylethynyl)anthracene
SMILES ClC1=C2C(=CC=C1)C(C#CC1=CC=CC=C1)=C1C=CC=CC1=C2C#CC1=CC=CC=C1

Specifications

Melting Point 203°C
Quantity 10 g
Formula Weight 412.92
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chloro-9,10-bis(phenylethynyl)anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.