Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Chloro-N,N,2-Trimethyl-1-Propenylamine

Catalog No. 4982705ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-827-05ML 5 mL
49-827-050ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 49-827-05ML Supplier Sigma Aldrich Supplier No. 4982705ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Chloro-N,N,2-trimethyl-1-propenylamine is an acid halogenation reagent developed by Ghosez, that enables the conversion of carboxylic acids into the corresponding chlorides under strictly neutral conditions. This method was successfully used in the total synthesis of caloporoside, roseophilin, (-)-enniatin B, (+/-)-epimeloscine and (+/-)-meloscine.

Specifications

CAS 26189-59-3
Density 1.01 g/mL at 25°C
Boiling Point 129°C to 130°C
Molecular Formula C6H12ClN
Refractive Index n20/D 1.453
Linear Formula (CH3)2C=C(Cl)N(CH3)2
MDL Number MFCD00800562
Quantity 5 mL
Synonym Ghosez's reagent
Molecular Weight (g/mol) 133.62 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.