Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloroadamantane 98.0+%, TCI America™
SDP

Catalog No. C074725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C074725G 25 g
C07475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C074725G Supplier TCI America Supplier No. C074725G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 935-56-8
Molecular Formula C10H15Cl
Molecular Weight (g/mol) 170.68
MDL Number MFCD00075627
InChI Key OZNXTQSXSHODFR-UHFFFAOYSA-N
Synonym 1-adamantyl chloride, adamantyl chloride, 3-chloroadamantane, adamantane, 1-chloro, tricyclo 3.3.1.13,7 decane, 1-chloro, 1-chlorotricyclo 3.3.1.13,7 decane, 3r,5s,7s-1-chloroadamantane, 1-chlorotricyclo 3.3.1.1∼3,7∼ decane
PubChem CID 64154
IUPAC Name 1-chloroadamantane
SMILES ClC12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 166°C
Color White
Quantity 25 g
Formula Weight 170.68
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chloroadamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.