Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorobutane, For Synthesis, MilliporeSigma™

Catalog No. M8016401000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M8016401000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M8016401000 Supplier MilliporeSigma Supplier No. 8.01640.1000
Add to Cart
Add to Cart

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Melting Point -123°C
Packaging Glass Bottle
Quantity 1 L
UN Number 1127
Formula Weight 92.57
Physical Form Liquid
Assay ≥99%
Chemical Name or Material 1-Chlorobutane
Recommended Storage Store at 15° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.