Learn More
Sigma Aldrich Fine Chemicals Biosciences 1-Chlorobutane suitable fo
Catalog No. 502898886
Encompass
1-Chlorobutane is an alkyl halide. It undergoes reaction with nanocrystalline MgO at 200-350C to afford butane isomers and MgCl2 via dehydrochlorination.
Catalog No. 50-289-8886 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 349582L
1-Chlorobutane is an alkyl halide. It undergoes reaction with nanocrystalline MgO at 200-350C to afford butane isomers and MgCl2 via dehydrochlorination.