Learn More
Sigma Aldrich Fine Chemicals Biosciences 1-Chlorobutane suitable fo1L
Catalog No. 502898037
Encompass
1-Chlorobutane is an alkyl halide. It undergoes reaction with nanocrystalline MgO at 200-350C to afford butane isomers and MgCl2 via dehydrochlorination.
Catalog No. 50-289-8037 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 349581L
1-Chlorobutane is an alkyl halide. It undergoes reaction with nanocrystalline MgO at 200-350C to afford butane isomers and MgCl2 via dehydrochlorination.