Learn More
Medchemexpress LLC 1-(chlorocarbonyl)piperidine-d10 | 1219803-31-2 | >98.0% | 10 MG
SDP
Catalog No. 5000296868
Encompass_Preferred
1-(Chlorocarbonyl)piperidine-d10 is a deuterium-labeled form of 1-(chlorocarbonyl)piperidine intended for use as an isotope-labeled tracer or internal standard in analytical, pharmacokinetic, and drug metabolism studies. The compound is a small-quantity solid with formula C6D10ClNO, molecular weight 157.66, and CAS 1219803-31-2, provided for research and analytical applications.
  • Deuterium-labeled tracer for quantitative mass spectrometry.
  • Provides a defined d10 isotopic shift for internal standardization.
  • Supplied as a small-quantity solid suitable for analytical workflows.
  • Useful in ADME and metabolic profiling applications.
  • Handle and store according to SDS recommendations for acid chloride derivatives.
Catalog No. 50-002-96868 Supplier Medchemexpress LLC Supplier No. HY33540S10MG
1-(Chlorocarbonyl)piperidine-d10 is a deuterium-labeled form of 1-(chlorocarbonyl)piperidine intended for use as an isotope-labeled tracer or internal standard in analytical, pharmacokinetic, and drug metabolism studies. The compound is a small-quantity solid with formula C6D10ClNO, molecular weight 157.66, and CAS 1219803-31-2, provided for research and analytical applications.
  • Deuterium-labeled tracer for quantitative mass spectrometry.
  • Provides a defined d10 isotopic shift for internal standardization.
  • Supplied as a small-quantity solid suitable for analytical workflows.
  • Useful in ADME and metabolic profiling applications.
  • Handle and store according to SDS recommendations for acid chloride derivatives.