Learn More
Medchemexpress LLC 1-(Chlorocarbonyl)piperidine-d10 | 1219803-31-2 | 98.2% | 157.66 | 1 MG
SDP
Catalog No. 5000404142
Encompass_Preferred
1-(Chlorocarbonyl)piperidine-d10 is the deuterium labeled 1-(Chlorocarbonyl)piperidine. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-04142 Supplier Medchemexpress LLC Supplier No. HY33540S1MG
1-(Chlorocarbonyl)piperidine-d10 is the deuterium labeled 1-(Chlorocarbonyl)piperidine. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.