Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Chloroethyl chloroformate

Catalog No. 3014855G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-148-55G 5 g
30-148-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-148-55G Supplier Sigma Aldrich Supplier No. 3014855G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-chloroethyl chloroformate was used:
  • For n-demethylation reaction during determination of multiple drugs of abuse in biological fluids using capillary electrophoresis with native fluorescence and laser-induced fluorescence detection
  • As catalyst in chemoselective desilylation of silyl-protected alcohols
  • As reagent in n-demethylation of tertiary amines to produce drug metabolite reference material for forensic toxicological applications
  • As reagent to cleave benzhydryl groups from amines

Specifications

CAS 50893-53-3
Density 1.325 g/mL (at 25°C (literature))
Boiling Point 118°C to 119°C (lit.)
Molecular Formula C3H4Cl2O2
Refractive Index n20/D 1.422 (literature)
Linear Formula CH3CHClOCOCl
MDL Number MFCD00000647
Quantity 5 g
Synonym ACE-Cl
Molecular Weight (g/mol) 142.97
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.