Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloroethyl Cyclohexyl Carbonate 98.0+%, TCI America™
SDP

Catalog No. C1786250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1786250G 250 g
C178625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1786250G Supplier TCI America Supplier No. C1786250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99464-83-2
Molecular Formula C9H15ClO3
Molecular Weight (g/mol) 206.666
MDL Number MFCD04038149
InChI Key ONZWFHWHTYZZLM-UHFFFAOYSA-N
Synonym Carbonic Acid 1-Chloroethyl Cyclohexyl Ester
PubChem CID 11229529
IUPAC Name 1-chloroethyl cyclohexyl carbonate
SMILES CC(OC(=O)OC1CCCCC1)Cl

Specifications

Color Yellow
Boiling Point 107°C
Quantity 250 g
UN Number 1760
Formula Weight 206.67
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloroethyl Cyclohexyl Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.