Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(Chloromethyl)naphthalene

Catalog No. 2517050ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-170-50ML 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-170-50ML Supplier Sigma Aldrich Supplier No. 2517050ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-(Chloromethyl)naphthalene

Specifications

CAS 86-52-2
Melting Point 32°C (lit.)
Density 1.18 g/mL (at 25°C (literature))
Boiling Point 167°C to 169°C (25 mmHg)
Molecular Formula C11H9Cl
Refractive Index n20/D 1.635 (literature); n20/D 1.635
Linear Formula C10H7CH2Cl
MDL Number MFCD00004042
Quantity 25 mg
Synonym 1-Naphthylmethyl chloride
Molecular Weight (g/mol) 176.64
Percent Purity ≥97.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.