Learn More
Medchemexpress LLC 1-Chloromethylpyrene 250mg
SDP
Catalog No. 5000205221
Encompass_Preferred
1-Chloromethylpyrene is an alkylating agent that enters the DNA double helix structure through intercalation forms covalent bonds with electrophilic groups resulting in DNA cross-linking thereby inhibiting the proliferation of tumor cells[1]
Catalog No. 50-002-05221 Supplier Medchemexpress LLC Supplier No. 250MGHYW111005
1-Chloromethylpyrene is an alkylating agent that enters the DNA double helix structure through intercalation forms covalent bonds with electrophilic groups resulting in DNA cross-linking thereby inhibiting the proliferation of tumor cells[1]