Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene

Catalog No. AC180970025
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC180970025 2.5 L Glass bottle
AC180971000 100 mL Glass bottle
AC180970010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC180970025 Supplier Thermo Scientific Chemicals Supplier No. 180970025
Only null left
Add to Cart

CAS: 90-13-1 | C10H7Cl | 162.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-13-1 , 91-58-7
Molecular Formula C10H7Cl
Molecular Weight (g/mol) 162.62
InChI Key JTPNRXUCIXHOKM-UHFFFAOYSA-N
Synonym alpha-chloronaphthalene, naphthalene, 1-chloro, 1-naphthyl chloride, chloronaphthalene, 1-chloro-naphthalene, alpha-naphthyl chloride, 1-chlornaftalen, 1-chloro naphthalene, alpha-chlornaphthalene, .alpha.-chloronaphthalene
PubChem CID 7003
IUPAC Name 1-chloronaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Cl

Specifications

Melting Point -2.5°C
Color Colorless to Yellow
Density 1.1940g/mL
Boiling Point 259.0°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.6310 to 1.6340
Linear Formula C10H7Cl
Quantity 2.5 L
Beilstein 05,541
Specific Gravity 1.194
Merck Index 15,2151
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene,petroleum,ether and alcohol
Formula Weight 162.62
Percent Purity 85%
Grade Technical
Physical Form Liquid
Chemical Name or Material 1-Chloronaphthalene
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 201-967-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.