Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloropinacolin 97.0+%, TCI America™
SDP

Catalog No. C1335500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1335500ML 500 mL
C133525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1335500ML Supplier TCI America Supplier No. C1335500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13547-70-1
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00035688
InChI Key ULSAJQMHTGKPIY-UHFFFAOYSA-N
Synonym 1-chloropinacolone, 1-chloro-3,3-dimethyl-2-butanone, chlorpinakolin, 2-butanone, 1-chloro-3,3-dimethyl, 1-monochloropinacoline, tert-butyl chloromethyl ketone, alpha-chloropinacolin, alpha-chloropinacoline, chloromethyl tert-butyl ketone, chloropinacoline
PubChem CID 83572
IUPAC Name 1-chloro-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)CCl

Specifications

Color Yellow
Boiling Point 173°C
Quantity 500 mL
Formula Weight 134.60
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloropinacolin
EINECSNumber (2)-3557
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.