Learn More
OHAUS 1 Clip Plate for 100 mL Flasks
Catalog No. 01804333 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-333 Shaker Attachment
1 options
Catalog No. 01-804-333 Supplier OHAUS Supplier No. 83041372
Add to Cart
Add to Cart
Metal clip plate to accommodate up to 4 100 mL Volumetric Flasks.
  • This Metal clip plate can be attached to the Magnetic Platform for use with the Roto-Shake Genie, Rotator Genie, Enviro-Genie or Incubator-Genie.
TRUSTED_SUSTAINABILITY
Specifications
Holds Volumetric Flask
Product Type Shaker Attachment