Learn More
OHAUS 1 Clip Plate
Catalog No. 01804395 Shop All OHAUS Products
Change view
Click to view available options
Holds:
3 Tubes
6 Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
01-804-395 6 Tubes
01-804-341 3 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-804-395 Supplier OHAUS Supplier No. 83041443
Add to Cart
Add to Cart
Single metal clip plate to accommodate up to 6 tubes and 3 tubes with a diameter of 15-17 mm and 28-30 mm respectively.
  • A maximum of 8 clip plates, 4 per side
  • It may be used with the Roto-Shake Genie, Rotator Genie, Enviro-Genie, or Incubator Genie.
TRUSTED_SUSTAINABILITY
Specifications
Holds 6 Tubes
Product Type Shaker Attachment
Diameter (Metric) 15 to 17 mm