Learn More
Grainger Scientific Industries Clip Plate, 12-Tube Capacity, 10-13 Mm, 1 ea
Catalog No. 502768863
Encompass_Preferred
The Genie Clip Plate is designed to securely hold up to twelve tubes measuring 10-13 mm in diameter during mixing, rocking, or incubating applications. This accessory provides stable, organized support for laboratory workflows involving culture tubes and sample vials, ensuring reliable positioning even during vigorous agitation.
  • Holds 12 tubes with 10-13 mm diameters
  • Integrates seamlessly with compatible platforms
  • Ensures stable and organized sample support
  • Reliable tube positioning under agitation
  • Easy to clean or decontaminate
Catalog No. 50-276-8863 Supplier Grainger Supplier No. 13R347
The Genie Clip Plate is designed to securely hold up to twelve tubes measuring 10-13 mm in diameter during mixing, rocking, or incubating applications. This accessory provides stable, organized support for laboratory workflows involving culture tubes and sample vials, ensuring reliable positioning even during vigorous agitation.
  • Holds 12 tubes with 10-13 mm diameters
  • Integrates seamlessly with compatible platforms
  • Ensures stable and organized sample support
  • Reliable tube positioning under agitation
  • Easy to clean or decontaminate