Learn More
Weber Scientific #1 Cover Glasses, 1oz, about 170 pieces
SDP
Catalog No. NC1632315
Encompass
This item has a minimum order quantity of 2 packs per supplier requirements
High quality optical glass. Standard 22 X 22 mm size. Thickness 0.13 to 0.17 mm. About 170 pieces per oz.
Catalog No. NC1632315 Supplier Weber Scientific Supplier No. 201225
This item has a minimum order quantity of 2 packs per supplier requirements
High quality optical glass. Standard 22 X 22 mm size. Thickness 0.13 to 0.17 mm. About 170 pieces per oz.