Learn More
Medchemexpress LLC HY-110095 10mg Medchemexpress, (±)-CPSI-1306 CAS:1309793-47-2 Purity:>98%
SDP
Catalog No. 501968743
Encompass_Preferred
Medchemexpress, HY-110095 10mg (±)-CPSI-1306 CAS:1309793-47-2 (±)-CPSI-1306 is an orally available antagonist of macrophage migration inhibitory factor (MIF). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8743 Supplier Medchemexpress LLC Supplier No. HY11009510MG
Medchemexpress, HY-110095 10mg (±)-CPSI-1306 CAS:1309793-47-2 (±)-CPSI-1306 is an orally available antagonist of macrophage migration inhibitory factor (MIF). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.