Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate, 98%

Catalog No. AAH59334ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59334ME 500 mg
AAH59334MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59334ME Supplier Thermo Scientific Chemicals Supplier No. H59334ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 59016-56-7 | C8H10BF4N3 | 234.99 g/mol

1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate is a cyanylation reagent for protein sulfhydryl groups and used to prepare protein-polysaccharide conjugates. It is employed as an activating agent for polysaccharide resins and conjugation of lipopolysaccharides while retaining endotoxic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate is a cyanylation reagent for protein sulfhydryl groups and used to prepare protein-polysaccharide conjugates. It is employed as an activating agent for polysaccharide resins and conjugation of lipopolysaccharides while retaining endotoxic activity.

Solubility
Soluble in water, and acetonitrile.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59016-56-7
Molecular Formula C8H10BF4N3
Molecular Weight (g/mol) 234.99
MDL Number MFCD00011998
InChI Key MBLVMDCQDCVKNE-UHFFFAOYSA-N
Synonym cdap, 1-cyano-4-dimethylamino pyridinium tetrafluoroborate, 1-cyano-4-dimethylamino pyridin-1-ium tetrafluoroborate, unii-p4w72066jt, 1-cyano-4-dimethylamino-pyridinium tetrafluoroborate, pubchem17735, acmc-209m9a, n-cyano-4-dimethylaminopyridinium tetrafluoroborate, 1-cyano-4-dimethylaminopyridinium tetra-fluoroborate, n-cyano-4-dimethylamino-pyridinium tetrafluoroborate
PubChem CID 9881151
IUPAC Name 4-(dimethylamino)pyridin-1-ium-1-carbonitrile;tetrafluoroborate
SMILES F[B-](F)(F)F.CN(C)C1=CC=[N+](C=C1)C#N

Specifications

Melting Point 196°C to 200°C
Quantity 500 mg
Beilstein 5685616
Solubility Information Soluble in water,and acetonitrile.
Formula Weight 234.99
Percent Purity 98%
Chemical Name or Material 1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.