Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclohexenylboronic acid, 97%

Catalog No. AC430180010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430180010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430180010 Supplier Thermo Scientific Chemicals Supplier No. 430180010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 89490-05-1 | C6H11BO2 | 125.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89490-05-1
Molecular Formula C6H11BO2
Molecular Weight (g/mol) 125.96
MDL Number MFCD02179497
InChI Key XZWQKJXJNKYMAP-UHFFFAOYSA-N
Synonym cyclohex-1-en-1-ylboronic acid, 1-cyclohexenylboronic acid, 1-cyclohexen-1-yl-boronic acid, cyclohexenylboronic acid, cyclohexen-1-yl-boronic acid, 1-cyclohexen-1-ylboronic acid, cyclohexene-1-boronic acid, 1-boronocyclohex-1-ene, cyclohex-1-enylboronic acid, cyclohexen-1-yl boronic acid
PubChem CID 3862902
IUPAC Name cyclohexen-1-ylboronic acid
SMILES OB(O)C1=CCCCC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 139.6°C to 140.3°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 125.96
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Cyclohexenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.