Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclohexyl-1-butanol 98.0+%, TCI America™
SDP

Catalog No. C15685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15685G Supplier TCI America Supplier No. C15685G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4352-42-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00019360
InChI Key MTUCYAOJXPTLHZ-UHFFFAOYNA-N
Synonym 1-cyclohexyl-1-butanol, cyclohexanemethanol, .alpha.-propyl, alpha-propylcyclohexanemethanol, zyklohexyl-butanol, cyclohexanemethanol, alpha-propyl, acmc-1ctfc, 1-hydroxybutyl cyclohexane, n-propyl cyclohexyl carbinol, mtucyaojxptlhz-uhfffaoysa
PubChem CID 107269
IUPAC Name 1-cyclohexylbutan-1-ol
SMILES CCCC(O)C1CCCCC1

Specifications

Color Colorless
Boiling Point 82°C
Quantity 5 g
Formula Weight 156.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Cyclohexyl-1-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.