Learn More
Medchemexpress LLC 1-cyclohexyl-3-dodecyl urea | 402939-18-8 | MFCD01344729 | 98.0% | 310.52 g/mol | C19H38N2O | 5 MG
SDP
Catalog No. 5000223423
Encompass_Preferred
1-Cyclohexyl-3-dodecyl urea is a small-molecule soluble epoxide hydrolase (sEH) inhibitor supplied as a white to off-white solid for research use. It is provided in milligram-scale packs for biochemical and pharmacological studies involving sEH activity.
  • Highly selective sEH inhibitor for research applications.
  • High purity (98.0%) suitable for in vitro studies.
  • Supplied as a white to off-white solid for straightforward handling.
  • Available in small milligram pack sizes for screening and assay work.
  • Molecular weight 310.52 g/mol and formula C19H38N2O for experimental planning.
Catalog No. 50-002-23423 Supplier Medchemexpress LLC Supplier No. HY1357955MG
1-Cyclohexyl-3-dodecyl urea is a small-molecule soluble epoxide hydrolase (sEH) inhibitor supplied as a white to off-white solid for research use. It is provided in milligram-scale packs for biochemical and pharmacological studies involving sEH activity.
  • Highly selective sEH inhibitor for research applications.
  • High purity (98.0%) suitable for in vitro studies.
  • Supplied as a white to off-white solid for straightforward handling.
  • Available in small milligram pack sizes for screening and assay work.
  • Molecular weight 310.52 g/mol and formula C19H38N2O for experimental planning.