Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-cyclohexyl-3-dodecylurea | 402939-18-8 | MFCD01344729 | 98.0% | C19H38N2O | 10MG
SDP

Catalog No. 5000259205
Encompass_Preferred

1-Cyclohexyl-3-dodecylurea is a small-molecule urea reported as a selective soluble epoxide hydrolase (sEH) inhibitor for research use. It is supplied as a solid reagent with high listed purity and is typically used in biochemical and pharmacological studies of sEH modulation.

  • High purity (98.0%).
  • Molecular formula C19H38N2O.
  • Molecular weight 310.52 g/mol.
  • Solid physical form for convenient handling and storage.
  • Offered in small research pack sizes (5-100 mg).
  • Used as a tool compound for sEH inhibition studies.

Catalog No. 50-002-59205 Supplier Medchemexpress LLC Supplier No. HY13579510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Cyclohexyl-3-dodecylurea is a small-molecule urea reported as a selective soluble epoxide hydrolase (sEH) inhibitor for research use. It is supplied as a solid reagent with high listed purity and is typically used in biochemical and pharmacological studies of sEH modulation.

  • High purity (98.0%).
  • Molecular formula C19H38N2O.
  • Molecular weight 310.52 g/mol.
  • Solid physical form for convenient handling and storage.
  • Offered in small research pack sizes (5-100 mg).
  • Used as a tool compound for sEH inhibition studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.