Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Cyclohexylcarbonyl)piperazine, 97%, Thermo Scientific™

Catalog No. AAH55480MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55480MD 250 mg
AAH5548003 1 g
AAH5548006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH55480MD Supplier Thermo Scientific Chemicals Supplier No. H55480MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27561-62-2
Molecular Formula C11H20N2O
Molecular Weight (g/mol) 196.294
MDL Number MFCD06798113
InChI Key ZSZROXCAFYZNHE-UHFFFAOYSA-N
Synonym 1-cyclohexylcarbonyl piperazine, cyclohexyl piperazin-1-yl methanone, 1-cyclohexanecarbonylpiperazine, methanone, cyclohexyl-1-piperazinyl, maybridge1_006754, cyclohexylpiperazino ketone, cyclohexyl piperazinyl ketone, n-cyclohexylcarbonyl piperazine, cyclohexyl-1-piperazinylmethanone, 1-cyclohexanecarbonyl-piperazine
PubChem CID 3437502
IUPAC Name cyclohexyl(piperazin-1-yl)methanone
SMILES C1CCC(CC1)C(=O)N2CCNCC2

Specifications

Melting Point 84°C to 89°C
Color White to Brown
Quantity 250 mg
Formula Weight 196.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(Cyclohexylcarbonyl)piperazine
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.