Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclohexylpiperidine 98.0+%, TCI America™
SDP

Catalog No. C30325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30325ML 5 mL
C303225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30325ML Supplier TCI America Supplier No. C30325ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3319-01-5
Molecular Formula C11H21N
Molecular Weight (g/mol) 167.30
MDL Number MFCD00014642
InChI Key DSSKLTAHHALFRW-UHFFFAOYSA-N
Synonym cyclohexylpiperidine, n-cyclohexylpiperidine, piperidine, 1-cyclohexyl, 1-piperidinocyclohexane, 1-cyclohexyl-piperidine, acmc-1cpdp, cyclohexyl piperidine, wln: t6ntj a-al6tj, 5-20-02-00040 beilstein handbook reference
PubChem CID 18724
ChEBI CHEBI:38897
IUPAC Name 1-cyclohexylpiperidine
SMILES C1CCC(CC1)N2CCCCC2

Specifications

Color Yellow
Boiling Point 107°C
Quantity 5 mL
Formula Weight 167.30
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Cyclohexylpiperidine
RTECSNumber TM6520000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.