Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclopentenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C28935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2893-5G 5 g
C28931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2893-5G Supplier TCI America Supplier No. C28935G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 850036-28-1
Molecular Formula C5H9BO2
Molecular Weight (g/mol) 111.935
MDL Number MFCD02179496
InChI Key UZBHNSVUMGIKLU-UHFFFAOYSA-N
PubChem CID 3861343
IUPAC Name cyclopenten-1-ylboronic acid
SMILES B(C1=CCCC1)(O)O

Specifications

Melting Point 140°C
Quantity 5 g
Synonym cyclopent-1-en-1-ylboronic acid, 1-cyclopentenylboronic acid, cyclopentenylboronic acid, cyclopent-1-en-1-yl boronic acid, cyclopentene-1-boronic acid
Formula Weight 111.94
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.