Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclopentylpiperazine 98.0+%, TCI America™
SDP

Catalog No. C24665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24665G Supplier TCI America Supplier No. C24665G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21043-40-3
Molecular Formula C9H18N2
Molecular Weight (g/mol) 154.257
MDL Number MFCD00168029
InChI Key PVMCQBPJKPMOKM-UHFFFAOYSA-N
PubChem CID 806421
IUPAC Name 1-cyclopentylpiperazine
SMILES C1CCC(C1)N2CCNCC2

Specifications

Color White
Boiling Point 105°C
Quantity 5 g
Formula Weight 154.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Cyclopentylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.